Would you like to log in again?
Nitrate/Nitrite Fluorometric Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
780051-192
75817-232EA 410.44 USD InStock
75817-232
Nitrate/Nitrite Fluorometric Assay Kit, Cayman Chemical Company
Assays Cellular Assays
For the quantitation of low levels of nitrate and nitrite in biological samples (particularly tissue culture medium).
  • Measure nitrate, nitrite, and/or total nitrate/nitrites in plasma, serum, tissue culture media, and tissue homogenates
  • Assay 40 samples in duplicate/96-well plate
  • Assay Range: 60 nM-3.85 μM (nitrate and/or nitrite)
  • Plate-based fluorometric measurement (ex 360-365 nm, em 430 nm)
Nitric Oxide (NO) is produced in trace quantities by neurons, endothelial cells, platelets, and neutrophils in response to homeostatic stimuli
Cayman Chemical’s Nitrate/Nitrite Fluorometric Assay Kit provides a convenient method for the quantitation of low levels of NO2- and NO3- in biological samples (particularly tissue culture medium). Previously undetectable concentrations of nitrate/nitrite in a variety of samples can be accurately quantitated using this kit as it extends the lower limit of detection by 20-fold over our popular colorimetric version. The minimum detectable quantity of NO2/NO3 is
50 nM.
This NO is scavenged rapidly (t½ = 4 s) and acts in a paracrine fashion to transduce cellular signals. NO interacts with the HEME prosthetic group of guanylate cyclase, activating the enzyme and leading to increased cGMP levels. Other cells (macrophages, fibroblasts, and hepatocytes) also produce NO in micromolar concentrations in response to inflammatory or mitogenic stimuli. These very high levels of NO lead to the formation of peroxynitrite, destruction of iron-sulphur clusters, thiol nitrosation, and nitration of protein tyrosine residues. Thus, the amount of NO produced in different biological systems can vary over several orders of magnitude and its subsequent chemical reactivity is diverse. The final products of NO in vivo are nitrite (NO2-) and nitrate (NO3-). Relative proportions of NO2- and NO3- within a sample are variable and cannot be predicted with certainty. Therefore, the best index of total NO production is the sum of both of these products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty