Would you like to log in again?
NOS Activity Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
781001-1
75817-234EA 428.25 USD InStock
75817-234
NOS Activity Assay Kit, Cayman Chemical Company
Assays Enzyme Assays
For the measurement of NOS activity.
  • Measure NOS activity in cell lysates and purified preparations
  • Material for 50 reactions
  • Radioactive substrate not supplied
The NOS Activity Assay measures NOS activity by monitoring the conversion of radiolabeled arginine to citrulline. This assay is simple, sensitive, and specific for NOS activity and can be used with both crude and purified enzyme preparations. The kit includes sufficient materials and reagents for 50 total reactions. Radiolabeled arginine and NADPH are not included with the kit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty