Would you like to log in again?
Prostaglandin D₂-MOX ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
512011-96 512011-480
75817-270EA 424.37 USD InStock
75817-270 75817-268
Prostaglandin D₂-MOX ELISA Kit, Cayman Chemical
Assays ELISAs
Competitive ELISA for quantitation of PGD2 in biological samples.
  • Treatment of samples with MOX hydrochloride converts unstable PGD2 to stable PGD2-MOX
  • Highly sensitive
  • Measure PGD2-MOX levels down to 3.1 pg/mL
  • Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
  • Midpoint: 50% B/B0: 10 – 35 pg/mL
This PGD2-MOX ELISA is based on the conversion of PGD2 to a stable MOX derivative. Treatment of the sample with MOX HCl converts PGD2 into PGD2-MOX, preventing its further chemical degradation. The antiserum used in the assay was developed using conjugates of this derivative and is very specific for PGD2-MOX.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty