Would you like to log in again?
Prostaglandin D₂ (PGD₂) ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical
512031-480 512031-96
75817-272EA 2162.02 USD InStock
75817-272 75817-274
Prostaglandin D₂ (PGD₂) ELISA Kit, Cayman Chemical
Assays ELISAs
A competitive assay that can be used for quantification of PGD₂.
  • Measure PGD₂ in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure PGD₂ levels down to 55 pg/ml
  • Incubation: 18 hours | Development: 90-120 minutes | Read: Colorimetric at 405-420 nm
Prostaglandin D2 (PGD₂) is biosynthesized in the brain by a soluble, 26 kDa glutathione-independent lipocalin-type PGD₂ synthase. This PGD₂ accumulates in the cerebrospinal fluid (CSF), where it induces physiologic sleep in rats and humans. PGD₂ is also synthesized in mast cells and leukocytes by a cellular, myeloid-type, glutathione-dependent PGD synthase. This PGD₂ which is formed in the intracellular and vascular compartments is rapidly metabolized to 11β-PGF and other metabolites. Measurement of the parent eicosanoid PGD₂ is appropriate in cell culture lysates and in CSF, where concentrations of several hundred pg/ml have been measured. Cayman’s PGD₂ ELISA Kit is a competitive assay that can be used for quantification of PGD₂ in cell culture lysates and enzymatic reactions.
Delivery information: Kit contains: Antibody, Tracer, Standard, Buffers, Reagents, and Plate
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty