Would you like to log in again?
DNA Methylation ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
589324-96 589324-480
75817-292EA 2125.6 USD InStock
75817-292 75817-294
DNA Methylation ELISA Kit, Cayman Chemical Company
Assays ELISAs
A competitive assay that can be used for the quantification of 5-methyl-2'-deoxy cytidine in urine, culture supernatants, plasma, and other sample matrices.
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure 5-methyl-2'-deoxy cytidine levels down to three ng/ml
  • Read: Colorimetric at 405 to 420 nm
DNA methylation is an important epigenetic process regulating gene expression. Methylation occurs on carbon 5 of 2'-deoxy-cytidine yielding the modified base 5-methyl-2'-deoxy cytidine. The methylation pattern of cells is tightly regulated during development with the methylation profile being transmitted from parent to daughter cells during cell division.
Methylation results in long-term silencing of genes, while unmethylated regions of DNA can be actively transcribed. Global changes in methylation can be quantified by measuring plasma or urinary levels of 5-methyl-2'-deoxy cytidine. These changes in methylation can provide valuable information about cancer status of an individual. For example, patients with leukemia excrete significantly elevated levels of 5-methyl-2'-deoxy cytidine compared to healthy individuals.
Global methylation within tissues can be measured in a similar manner, allowing study of tissue-specific changes that occur as a result of differentiation, aging, or carcinogenesis. Cayman’s DNA Methylation ELISA Kit is a competitive assay that can be used for the quantification of 5-methyl-2'-deoxy cytidine in urine, culture supernatants, plasma, and other sample matrices.
Delivery information: Kit contains antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, ellman's reagent, tracer and antiserum dye.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty