Would you like to log in again?
Superoxide Dismutase Assay Kit, Cayman Chemical
Supplier: Cayman Chemical
706002-96 706002-480
75817-336EA 777.37 USD InStock
75817-336 75817-338
Superoxide Dismutase Assay Kit, Cayman Chemical
Assays Enzyme Assays
For the measurement of SOD activity from plasma, serum, tissue homogenates, and cell lysates.
  • Measure copper/zinc, manganese, and iron SOD in tissues, plasma, serum, and cell lysates
  • Assay 41 samples in duplicate
  • Measure SOD activity down to 0.005 U/ml
Significant amounts of superoxide dismutase (SOD) in cellular and extracellular environments are crucial for the prevention of diseases linked to oxidative stress
Each kit contains sufficient reagents to assay 41 samples in duplicate and includes assay buffer, sample buffer, radical detector, SOD (standard), xanthine oxidase, a 96 well plate, and complete instructions.
Mutations in SOD account for approximately 20% of familial amyotrophic lateral sclerosis (ALS) cases. SOD also appears to be important in the prevention of other neurodegenerative disorders such as Alzheimer’s, Parkinson’s, and Huntington’s Diseases. The reaction catalyzed by SOD is extremely fast, having a turnover of 2 x 106 M−1sec−1 and the presence of sufficient amounts of the enzyme in cells and tissues typically keeps the concentration of superoxide very low. Quantification of SOD activity is therefore essential in order to fully characterize the antioxidant capabilities of a biological system. The Cayman Chemical SOD Assay kit is a fast and reliable assay for the measurement of SOD activity from plasma, serum, tissue homogenates, and cell lysates. SOD activity is assessed by measuring the dismutation of superoxide radicals generated by xanthine oxidase and hypoxanthine in a convenient 96 well format. The standard curve generated using this enzyme provides a means to accurately quantify the activity of all three types of SOD (Cu/Zn, Mn, and FeSOD).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty