Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclic GMP ELISA Kit, Cayman Chemical Company

Supplier: Cayman Chemical
581021-480 581021-96
75817-370EA 1670.85 USD InStock
75817-370 75817-372
Cyclic GMP ELISA Kit, Cayman Chemical Company
Assays ELISAs
For quantification of cGMP directly obtained from cell lysates, tissue homogenates, plasma, or urine.

  • Detect non-acetylated or acetylated cGMP in cell lysates, tissue homogenates, plasma, or urine
  • Measure non-acetylated cGMP levels down to 1 pmol/ml or acetylated cGMP levels down to 0.1 pmol/ml
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Contains acetic anhydride
  • Incubation: 18 hours | Development: 60-90 minutes | Read: Colorimetric at 405-420 nm

The Cayman Chemical cGMP Assay is a competitive ELISA that can be used for quantification of cGMP directly obtained from cell lysates, tissue homogenates, plasma, or urine. The assay has a range from 0.23-30 pmol/ml and a sensitivity (80% B/B0) of approximately 1 pmol/ml. Since the antibody used in this assay was prepared against a cGMP-carrier protein conjugate, antibody binding is increased if an acetyl group is present on the 2' hydroxyl group of the cGMP. The optional acetylation procedure for both samples and standards increases the sensitivity of the assay approximately 10 fold. A protocol for acetylating both the standards and samples prior to performing the assay is provided. Basal levels of cGMP in cell lysates can often be measured without acetylation, but results will depend on the type and number of cells being utilized. Platelets produce approximately 1.5-2.5 pmol cGMP/109 platelets under basal conditions. Cells such as NG108-15 cells and monocytes produce considerably more cGMP than platelets (approximately 0.1-1 pmol/106 cells).

Delivery information: Kit contains: Antiserum, tracer, standard, buffers, reagents, and plate(s).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty