Would you like to log in again?
Vannas Micro Scissors with Bayonet Handle, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
98-7936 98-7950 98-7952
10185-198EA 3105.93 USD InStock
10185-198 75818-106 75818-104
Vannas Micro Scissors with Bayonet Handle, Sklar
Scissors
Sklar's® Vannas Micro Scissors are most commonly used in iridectomy, cataract, and capsulotomy procedures.
  • Bayonet style handle
  • Reusable
  • Sterile
These double spring scissors are available straight or curved and in various lengths. The blades of these scissors are very small and sharp, being designed for use on specific sections of the eye. The micro blade can also be seen clearly under a microscope when used as a component of surgery.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty