Would you like to log in again?
BCYE Agar (Buffered Charcoal Yeast Extract), Hardy Diagnostics
Supplier: Hardy Diagnostics
G07
75820-342PK 170.25 USD InStock
75820-342
BCYE Agar (Buffered Charcoal Yeast Extract), Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of Legionella spp. from clinical and environmental specimens.
For the cultivation of Legionella spp
Buffered Charcoal Yeast Extract Agar For the cultivation of Legionella spp. from clinical and environmental specimens.
Conforms to ISO11371 formulation.
from clinical and environmental specimens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty