Would you like to log in again?
VIAL RND BTM CLR 22X38MM 20MM CRIMP 6ML
Supplier: Chemglass
Vial Clear Round Bottom 6.0mL^ Vial, Clear, Material: manufactured from Type 1 Class A Borosilicate glass, 20mm Headspace, Round Bottom, Dimensions: 22x38mm, 20mm Crimp, for use with Perkin Elmer Autosamplers, Size: 6.0mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL RND BTM CLR 22X38MM 20MM CRIMP 6ML 75820-648 CV-1281-2238 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty