Would you like to log in again?
INSERT FLAT BOTTOM GLASS 5X31MM 0.25ML
Supplier: Chemglass
Insert Glass 0.25mL^ Insert, Material: Glass, Flat Bottom, for opening of standard opening screw thread and standard opening crimp top vials containing a nominal I.D. Of 5mm, conical design permits complete sample removal, Size: 0.25mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INSERT FLAT BOTTOM GLASS 5X31MM 0.25ML 75820-690 CV-1414-0531 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty