Would you like to log in again?
INSERT GLASS BM WITH GLASS FLANGE 0.25ML
Supplier: Chemglass
Insert, 0.25mL, Glass, BM with Glass Flange. Both glass and plastic inserts have been engineered to fit the large opening in vials containing a nominal I.D. Of 6mm. Inserts are available assembled with a polymer bottom spring or patented Top Spring. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INSERT GLASS BM WITH GLASS FLANGE 0.25ML 75820-696 CV-1421-0631 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty