Would you like to log in again?
SEAL POLY CRIMP PTFE/SLN SEPTUM GR 11MM
Supplier: Chemglass
Poly Crimp* Seal Green 11mm^ Poly Crimp* Seal, Color: Green, PTFE/Silicone Septum, Material: Plastic, fits virtually any 11mm crimp sample vial, also permits an airtight seal by hand without crimping, better performance, Size: 11mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SEAL POLY CRIMP PTFE/SLN SEPTUM GR 11MM 75820-938 CV-3930-G011 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty