Would you like to log in again?
KIT VL/CLSR GPI 10-425 AMB 12X32MM 2ML
Supplier: Chemglass
Kit Srew Thread Vials and Closures^ Kit, Preassembled Large Opening Srew Thread Vials and Closures, Conatins: Amber 2.0mL 12x32 mm GPI 10-425 Vial and 0.010 in PTFE Septa Lined Closure, Pack of 100
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT VL/CLSR GPI 10-425 AMB 12X32MM 2ML 75820-974 CV-5015-1232 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty