Would you like to log in again?
Sterileze® Gas Sterilization Pouches, Amcor Flexibles
Supplier: Amcor Flexibles
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sterileze®
027712C 027714B 027711A 027713A
11213-268CS 2661.8 USD InStock
11213-268 11213-267 11213-265 75829-196
Sterileze® Gas Sterilization Pouches, Amcor Flexibles
Bags Sterilization Pouches
These clear plastic pouches feature a breathable Tyvek® vent that prevents blowup during sterilization cycles and allows fast aeration.
The Tower-Tear® snap-open feature provides quick, easy access to pouch contents. The pouches may be heat-sealed or taped.
Moisture and puncture-resistant pouches offer an excellent bacterial barrier. Pouches are 3 mil thick.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty