Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide 3% USP antiseptic 10 Volume, Hydrox Laboratories

Supplier: Healthcare Products

HDXD0012 HDXD0011
75830-256CS 38.65 USD InStock
75830-256 75830-254
Hydrogen peroxide 3% USP antiseptic 10 Volume, Hydrox Laboratories
Hydrogen peroxide


Formula: H₂O₂
MW: 34.01 g/mol
MDL Number: MFCD00011333
CAS Number: 7722-84-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty