Saint-Gobain manufactures PFA fittings for handling corrosive or ultra-pure fluids under contamination-free conditions.
- Simple design for ease of installation
- No special tools required
- Accommodates temperatures up to 212 °F (100 °C)
The PharmaFluor® compression fitting line offers PFA purity combined with the robust performance of compression style fittings.
We use 100% virgin HP PFA to give you the ultimate in purity and the minimum in contamination. Each PharmaFluor® PFA fitting is carefully inspected to ensure quality and reliability in your application and throughout the life of the product. Fluid handling applications in the biomedical, chemical, petrochemical, food processing and semiconductor industries benefit greatly from the use of these fittings.
Whether it be a simple fluid handling system or a complex laboratory layout, PharmaFluor® PFA fittings save precious time and money, and provide all the reliability needed to facilitate successful outcomes.
These fittings are ideal for gas or pneumatic connections in applications that do not require flared connections. Once tubing has been cut to the desired length, PharmaFluor® fittings require no tools for installation. The fittings are comprised of a high-purity PFA body, PVDF gripper, PTFE ferrule, and ETFE nut and are available in 1/8", 1/4", 3/8", 1/2", 3/4" and 1" sizes.