Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pharmafluor® FEP Tubing Connectors, Saint-Gobain Performance Plastics

Supplier: Saint-Gobain

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

G1077386 G1077063 G1077013 G1077090 G1077007 G1077053 G1077390 G1077019 G1077038 G1077062 G1077008 G1077091 G1077003 G1077088 G1077073 G1077382 G1077025 G1077388 G1077101 G1077057 G1077094 G1077005 G1077302 G1077377 G1077022 G1077026 G1077070 G1077024 G1077006 G1077066 G1077020 G1077103 G1077058 G1077012 G1077097 G1077001 G1077317 G1077333 G1077055 G1077071 G1077104 G1077067 G1077014 G1077316 G1077028 G1077086 G1077035 G1077318 G1077023 G1077029 G1077050 G1077321 G1077392 G1077036 G1077384 G1077395 G1077010 G1077056 G1077051 G1077004 G1077080 G1077021 G1077039 G1077037 G1077334 G1077002 G1077061 G1077338 G1077325 G1077060 G1077059 G1077009 G1077054 G1077027 G1077087 G1077015 G1077081 G1077096 G1077047 G1077072 G1077303 G1077068 G1077095 G1077322 G1077387
75830-408EA 137.09 USD InStock
75830-408 75830-528 75830-406 75830-526 75830-404 75830-448 75830-524 75830-402 75830-446 75830-456 75830-412 75830-532 75830-454 75830-498 75830-410 75830-530 75830-496 75830-452 75830-494 75830-450 75830-492 75830-490 75830-958 75830-518 75830-516 75830-956 75830-438 75830-954 75830-514 75830-436 75830-952 75830-512 75830-358 75830-522 75830-488 75830-400 75830-444 75830-960 75830-520 75830-442 75830-486 75830-484 75830-440 75830-482 75830-360 75830-480 75830-948 75830-508 75830-946 75830-506 75830-428 75830-944 75830-504 75830-426 75830-502 75830-942 75830-424 75830-468 75830-434 75830-478 75830-510 75830-356 75830-950 75830-476 75830-432 75830-398 75830-354 75830-430 75830-472 75830-470 75830-938 75830-418 75830-538 75830-416 75830-536 75830-414 75830-534 75830-940 75830-500 75830-422 75830-466 75830-420 75830-464 75830-462 75830-460
Pharmafluor® FEP Tubing Connectors, Saint-Gobain Performance Plastics
Fittings and Connectors
Saint-Gobain manufactures PFA fittings for handling corrosive or ultra-pure fluids under contamination-free conditions.

  • Simple design for ease of installation
  • No special tools required
  • Accommodates temperatures up to 212 °F (100 °C)

The PharmaFluor® compression fitting line offers PFA purity combined with the robust performance of compression style fittings.

We use 100% virgin HP PFA to give you the ultimate in purity and the minimum in contamination. Each PharmaFluor® PFA fitting is carefully inspected to ensure quality and reliability in your application and throughout the life of the product. Fluid handling applications in the biomedical, chemical, petrochemical, food processing and semiconductor industries benefit greatly from the use of these fittings.

Whether it be a simple fluid handling system or a complex laboratory layout, PharmaFluor® PFA fittings save precious time and money, and provide all the reliability needed to facilitate successful outcomes.

These fittings are ideal for gas or pneumatic connections in applications that do not require flared connections. Once tubing has been cut to the desired length, PharmaFluor® fittings require no tools for installation. The fittings are comprised of a high-purity PFA body, PVDF gripper, PTFE ferrule, and ETFE nut and are available in 1/8", 1/4", 3/8", 1/2", 3/4" and 1" sizes.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty