Would you like to log in again?
96-Well Polypropylene PCR Microplates, Greiner Bio-One
Supplier: Greiner Bio-One
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
652201 652270
75830-566CS 456.75 USD InStock
75830-566 82050-696
96-Well Polypropylene PCR Microplates, Greiner Bio-One
Microplates PCR Plates
These ultrathin polypropylene PCR plates are available with full, half, or no skirt to facilitate compatibility with a wide range of thermocyclers.
  • Polypropylene construction
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Available in a variety of skirt options
Plates are RNase-, DNase-, and pyrogen-free, and also free of human DNA.
Ordering information: Half skirt plate 82050-698 is compatible with ABI Sequencing Systems. Half skirt plate 82050-704 is compatible with the ABI PRISM® 7700.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty