Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pure-Fit® TCL Large Tube Clamps, Saint-Gobain Performance Plastics
Supplier: Saint-Gobain
PFTCL150 PFTCL250
75831-060BG 1497.8 USD InStock
75831-060 75831-062
Pure-Fit® TCL Large Tube Clamps, Saint-Gobain Performance Plastics
Clamps Tubing and Sanitary Clamps
Pure-Fit® TCL large tube clamps are designed to accommodate large diameter and thick wall tubing applications. They are compatible with standard, braided (reinforced) and multilayer tubing. Smooth edges and corners help prevent punctures and ruptures.
  • Provides complete fluid stoppage and can be utilized in the flow position
  • Features an integral bore hole for absolute lock-out capabilities
  • Can be installed on or over tubing or finished assemblies
  • High degree of leverage accommodates tubing with different durometers
  • Fully autoclavable and sterilizable
Constructed from animal derivative-free and BPA-free PVDF and available in two sizes to accommodate a wide range of thick wall tubing.
Areas of use include biopharmaceutical manufacturing, pharmaceutical processes, peristaltic pump sets, drug delivery & discovery, medical systems, laboratory functions and assemblies and tubing sets.
Certifications: Meets all USP Class VI criteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty