Would you like to log in again?
Armor Plate™ Milk Protein Coat Block and Diluent, ImmunO4
Supplier: ImmunO4
APMPC APMPC.1 APMPC.2
75831-570EA 84.36 USD InStock
75831-570 75831-942 75831-568
Armor Plate™ Milk Protein Coat Block and Diluent, ImmunO4
Protein Purification Protein Purification Reagents
Blocks and stabilizes coated ELISA plates and other solid phase substrates.
  • No Bovine serum proteins
  • Provides structural support and crowding agents
  • Minimizes background interactions and issues
  • Eliminates BSE regulatory concerns
  • ImmunO4 APMPC™ is compatible and works well in combination with ImmunO4 Stabilizers
  • Room temperature stable reagent
  • Stable through freeze cycles
  • Superior coat blocking
APMPC™ is a superior diluent that blocks, seals, and stabilizes with a consistent coat
APMPC™ can be used for antigens or antibodies. APMPC™ can be used for blocking and as a diluent buffer. An additional blocking step may not be required, potentially enabling blocking and stabilization in one step. Coated plates dried with APMPC™ have a long term shelf life, even at room temperature, when stored in a cool and dry environment. Antimicrobial agents are included to retard bacterial growth. APMPC™ should be tested for efficacy and stability in any new assay system.
This Coat Block was developed to substantially reduce Non Specific Antibody Binding and eliminate non-specific protein interactions. It is supplied as a one component liquid, ready-to-use formula.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty