Would you like to log in again?
Super Shield™ Peroxidase (HRP) Conjugate and Protein Stabilizer, ImmunO4
Supplier: ImmunO4
HRPZR HRPZR.2 HRPZR.1
75831-592EA 85.39 USD InStock
75831-592 75831-590 75831-588
Super Shield™ Peroxidase (HRP) Conjugate and Protein Stabilizer, ImmunO4
Protein Purification Protein Purification Reagents
Preserves and stabilizes the activity of HRP conjugates and proteins used in immunoassays and molecular assays.
  • No Mammalian proteins
  • Minimizes background interactions and issues
  • Eliminates BSE regulatory concerns
  • ImmunO4 HRPZR™ synergizes with ImmunO4 Armor Plate™ Non-Mammalian Protein Coat Block and Diluent to further improve unwanted non-specific binding
  • Room temperature stable reagent
  • Stable through freeze cycles
  • Superior HRP and protein stabilizer
Preserves and stabilizes the activity of HRP conjugates and proteins used in immunoassays and molecular assays. This stabilizer was developed to maximize the life of your HRP conjugates and proteins while eliminating non-specific protein interactions. It provides superior protection compared to competitor products. The stabilizer is supplied as a one component liquid ready to use formula. HRPZR™ is designed as a ready-to-use buffer and stabilizer for immunoassays and molecular assays. It can be used for high concentration stock and dilute conjugate solutions. Super Shield™ can be used directly as an assay buffer. Super Shield™ is stable and can be used at room temperature or 2° to 8°C. It stabilizes dilute conjugates for months at room temperature.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty