Would you like to log in again?
Super Shield™ Symulator™ Peroxidase (HRP) Conjugate and Protein Stabilizer, ImmunO4
Supplier: ImmunO4
SSSZR.1 SSSZR.2 SSSZR
75831-598EA 85.39 USD InStock
75831-598 75831-596 75831-594
Super Shield™ Symulator™ Peroxidase (HRP) Conjugate and Protein Stabilizer, ImmunO4
Protein Purification Protein Purification Reagents
Preserves and stabilizes the activity of Peroxidase (HRP) conjugates and proteins used in immunoassays and molecular assays.
  • Protein free
  • Uses natural biomolecular stabilizing agents
  • Can use as a Synthetic Replacement
  • Minimizes background interactions and issues
  • Eliminates BSE regulatory concerns
  • ImmunO4 SSSZR™ is compatible and works well with ImmunO4 Coat Block Diluents
  • Room temperature stable reagent
  • Stable through freeze cycles
  • Superior HRP and protein stabilizer
SSSZR™ is designed as a ready-to-use buffer and stabilizer for immunoassays and molecular assays
This protein-free stabilizer was developed with natural biomolecular stabilizing agents to maximize the life of your HRP conjugates and proteins while eliminating non-specific protein interactions. It provides superior protection compared to competitor products. The stabilizer is supplied as a one component liquid, ready-to-use formula.
It can be used for high concentration stock and dilute conjugate solutions. Super Shield™ can be used directly as an assay buffer. Super Shield™ is stable and can be used at room temperature or 2° to 8°C. It stabilizes dilute conjugates for months at room temperature. 
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty