Would you like to log in again?
Nalgene® Certified Clean Polycarbonate Biotainer™ Carboys, Sterile, Thermo Scientific
Supplier: Thermo Scientific
383423-42 383405-42 383405-16 383410-08 383410-42
75832-264CS 695.04 USD InStock
75832-264 75832-266 75832-272 75832-270 75832-268
Nalgene® Certified Clean Polycarbonate Biotainer™ Carboys, Sterile, Thermo Scientific
Bottles Carboys
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific Nalgene Polycarbonate Biotainer Bottles and Carboys reduce the risk of carry-over contamination.
  • Sterile and ready to use
  • Translucent construction
  • Robust and versatile
Durable, translucent PC construction ensures the integrity of your sample, even frozen to −100 °C. Sterile, ready-to-use products with a Sterility Assurance Level of 10⁻⁶ inside the bottle/closure. Robust, PC can be used from −100 to 100 °C, allowing for both heated mixing and frozen storage or shipment. Must be tested in actual use conditions as results may vary depending upon application. Inspection procedures throughout manufacturing include leak testing, dimensional testing, visual inspection for particulates and release testing to ensure the product meets all specifications. All container/closure systems are certified leakproof to protect valuable contents.
Certifications: Certified clean products, which meet USP <788> standards and endotoxins below 0.5 EU/ml.
Ordering information: Products can be supplied with forced extraction studies, validation binders and certificates.
Additionally our certified clean products include clean room produced in ISO Class 7 to limit particulates during molding; two heat-sealed sterile barriers to protect product prior to use; heat-sealed bags include quick-tears on the top and bottom and placed in carton liner twisted and tape/tie sealed to prevent gross contamination on the inner packaging; certified and lot-to-lot tested to meet USP <788> low particulate requirements to limit potential particulate contamination; certified low endotoxin (0.50 EU/ml) according to USP <85> to reduce pyrogens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty