Would you like to log in again?
VWR REFRIG 2 DOOR 55X30X79 IN 49 CU FT
Supplier: VWR International
Reach-In Refrigerator 55 x 30 x 79 in^ Reach-In Refrigerator, Door Type: Solid, Sections: 2, Door Style: Swing, 6 adjustable PVC-coated wire shelves, Interior lighting, 4in castors, Aluminum interior with stainless steel floor, R290 Hydrocarbon refrigerant, Capacity: 49 CU FT, Dimension: 55x30x79 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR REFRIG 2 DOOR 55X30X79 IN 49 CU FT 75832-274 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty