Would you like to log in again?
Trespa® Pegboards, AGR Fabricators
Supplier: AGR Fabricators
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AGPB-4836T-TR AGPB-3636-TR AGPB-2424T-TR AGPB-2430T-TR AGPB-3230-TR AGPB-3229-TR AGPB-4830-TR AGPB-3036T-TR AGPB-4230T-TR AGPB-2430-TR AGPB-2424-TR AGPB-1824-TR AGPB-2412T-TR AGPB-1830-TR AGPB-1830T-TR AGPB-3229T-TR AGPB-3230T-TR AGPB-3636T-TR AGPB-4830T-TR AGPB-2436-TR AGPB-4230-TR AGPB-2436T-TR AGPB-3030T-TR AGPB-1824T-TR AGPB-2030T-TR AGPB-3030-TR AGPB-2412-TR AGPB-4836-TR AGPB-3036-TR AGPB-2029-TR AGPB-2030-TR AGPB-2029T-TR
75832-316EA 350.61 USD InStock
75832-316 75832-318 75832-356 75832-378 75832-358 75832-386 75832-364 75832-366 75832-388 75832-382 75832-360 75832-384 75832-362 75832-380 75832-400 75832-368 75832-346 75832-348 75832-352 75832-396 75832-374 75832-452 75832-354 75832-376 75832-398 75832-370 75832-392 75832-394 75832-350 75832-372 75832-450 75832-390
Trespa® Pegboards, AGR Fabricators
Racks Draining Racks
Pegboards are made with durable Trespa® resin panels and can come with or without troughs.
  • Stainless steel or phenolic troughs
  • Trespa® resin construction
  • Durable
  • Available in a wide variety of size formats
  • Number of pegs varies between models
  • Lightweight
Pegboards are black in color, and 4" deep stainless steel or phenolic troughs are available if needed. Custom pegboards can be created if a different size is required.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty