Would you like to log in again?
CRITERION™ Dehydrated Culture Media, Barney Miller Broth with Indicator, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C8992
75832-478EA 720.25 USD InStock
75832-478
CRITERION™ Dehydrated Culture Media, Barney Miller Broth with Indicator, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Hardy Diagnostics CRITERION™ Barney Miller Broth with Indicator is recommended for the isolation and detection of beer spoilage microorganisms, specifically lactic acid bacteria
  • Wide mouth opening reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip enables 'no slip' finger holds
  • Stackable containers for efficient storage
  • Increased solubility; dissolves quickly
  • Custom and bulk products available to meet customer specifications
  • Complete technical/instruction sheets, available in HUGO software
  • Unparalleled performance; tested for optimal growth
CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards. This broth is supplied in a 2kg bucket with convenient screw lid.
Indicator allows for early detection of growth by yellow color change due to pH shift.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty