Would you like to log in again?
Annexin V Conjugates
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
29001 20128 AB287804-96TEST 29012 29069 29008 29005 29010 29057-100UL 29011 29085 99902 29014 29045-500UL 29013 29003 29045-100UL 29057-500UL 29009 29004
89138-864EA 393.54 USD InStock
89138-864 89138-854 76211-392 89410-660 89260-206 89138-850 89138-872 89138-660 89260-204 76414-940 89138-870 89410-658 75832-700 76414-938 76458-728 89138-868 89138-858 89411-114 89138-866 89138-856
Annexin V Conjugates
Protein Linking and Labeling Systems
Annexin V fluorescent conjugates can be used to detect apoptotic cells. Annexin V is available conjugated to a wide variety of dyes.
  • Detect apoptotic cells with fast, reliable detection of phosphatidylserine externalization
  • Conjugates with wide choice of CF® dye colors, plus biotin, R-PE, APC, and other fluorescent dyes
  • Biotium's CF® dyes have superior brightness, photostability, and are water-soluble
  • Can be used for fluorescence microscopy, live cell imaging, and flow cytometry
Annexin V fluorescent conjugates can be used to detect apoptotic cells. The human anticoagulant Annexin V is a 35 to 36 kDa calcium-dependent phospholipid-binding protein with high affinity for phosphatidylserine (PS). In normal viable cells, PS is located on the inner leaflet of the cytoplasmic membrane. In apoptotic cells, however, PS is translocated from the inner to the outer leaflet of the plasma membrane, where it can be detected by fluorescently labeled Annexin V.
Biotium’s next-generation CF® dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty