Would you like to log in again?
Campy, Blood Free, Karmali Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
G06
75832-740PK 60.36 USD InStock
75832-740
Campy, Blood Free, Karmali Agar, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Campy, Blood Free, Karmali Agar for Campylobacter contains charcoal and CVA antibiotics.
Karmali formula, with charcoal, deoxycholate, cefoperazone, vancomycin, and amphotericin. Plates are designed for the selective isolation of Campylobacter spp. The Karmali formula has been shown to achieve superior results in the recovery of Campylobacter from stool specimens. Also known as mCCDA (Modified Charcoal Cefoperazone Deoxycholate Agar and Campy CSM).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty