Would you like to log in again?
Protein Disulfide Isomerases (PDI) Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB284515-100TEST
75832-890EA 1256.39 USD InStock
75832-890
Protein Disulfide Isomerases (PDI) Inhibitor Screening Kit (Fluorometric), BioVision
Assays Protein Assays
Simple, HTP compatible assay to screen/characterize Protein Disulphide Isomerase (PDI) inhibitors within one hour.
  • A simple and high-throughput adaptable assay kit
  • Results within one hour
  • PDI converts Insulin into its reduced form
Protein Disulfide Isomerases (PDI, EC: 5.3.4.1) constitute a thio-disulfide oxidoreductase family. PDI is abundant in the lumen of endoplasmic reticulum (ER). PDI plays an essential role in catalyzing the rearrangement of S-S bonds in proteins and functions as a chaperon. Recent studies show that PDI activity is essential for cancer cell survival and proliferation, and targeting the PDI activity with its inhibitors abrogates survival responses to ER stress in cancer cells. Thus, identification and development of PDI inhibitors represents an important approach in cancer therapy. In BioVision’s PDI Inhibitor Screening Kit, the PDI converts Insulin into its reduced form in the presence of Dithiothreitol (DTT). This reduced insulin binds to a fluorescent probe to generate an intensely fluorescent product (Ex/Em = 440/490 nm). In the presence of a PDI inhibitor, the reaction is impeded/abolished, resulting in a decrease or total loss of fluorescence. This assay kit can be used to screen/study/characterize the potential inhibitors of Protein Disulfide Isomerase. The assay is simple, high-throughput adaptable and can be performed within one hour.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty