Would you like to log in again?
TEV Protease Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB211109-100TEST
75832-892EA 841.41 USD InStock
75832-892
TEV Protease Activity Assay Kit (Fluorometric), BioVision
Assays Protein Assays
Simple kit with a fast protocol to detect as low as 50 ng/well of TEV Protease activity in viral lysates and purified protein samples.
  • Simple kit
  • Fast protocol fluorometrically detects TEV Protease activity
TEV Protease (EC: 3.4.22.44, Tobacco Etch Virus protease) is a cysteine protease that recognizes the cleavage site of Glu-Xaa- Xaa-Y- Xaa-Gln-(Gly/Ser) and cleaves between Gln and Gly/Ser. The optimal sequence is Glu-Asn-Leu-Tyr-Phe-Gln-Ser/Glycine (ENLYFQS/G). TEV Protease has high specificity and great stability and is active over a wide range of temperatures (4-37°C) with an optimal activity at 34°C. BioVision’s TEV Protease Activity assay kit utilizes the ability of TEV Protease to cleave a synthetic Fluorescein-based peptide substrate to release fluorescein which can be easily quantified using a fluorometer or fluorescence microplate reader (Ex/Em = 490/560 nm). This assay kit is simple, rapid and can detect TEV Protease activity as low as 50 ng/well in viral lysates and purified protein samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty