Would you like to log in again?
PLTP Activity Fluorometric Assay Kit II, BioVision Inc.
Supplier: BioVision
AB196998-100TEST
75832-912EA 1248.72 USD InStock
75832-912
PLTP Activity Fluorometric Assay Kit II, BioVision Inc.
Assays Protein Assays
Measurement of PLTP activity in animal serum, plasma and recombinant protein.
  • Rabbit serum provided as a positive control
  • Uses a self-quenched fluorescent phospholipid
  • Can be measured when transferred to an acceptor molecule
Phospholipid transfer protein (PLTP) plays an important role in transferring phospholipids between HDL molecules to modulate HDL size and composition and controlling plasma HDL levels. PLTP also transfers phospholipids from lipoproteins to HDL. PLTP plays a key role in reverse cholesterol transport and may promote atherosclerosis. PLTP is, therefore, considered a promising target for pharmacological intervention. BioVision’s PLTP Activity Fluorometric Assay Kit II uses a self-quenched fluorescent phospholipid that can be measured when transferred to an acceptor molecule. The fluorometric intensity is directly proportional to the amount of phospholipid transferred. Rabbit serum is provided as a positive control. This Assay Kit, in addition to measuring activity in serum, is also suitable for testing activity of recombinant protein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty