Would you like to log in again?
Precision Count Beads™, Biolegend®
Supplier: BioLegend
424902 424902
75833-874EA 335.53 USD InStock
75833-874 CA75833-874
Precision Count Beads™, Biolegend®
Assays Cellular Assays
Precision Count Beads™ are fluorescent particles about 10 µm that can be detected across a broad range of wavelengths (405 - 635 nm excitation and 400 - 800 nm emission). Absolute numbers of cells can be determined by comparing the bead count and cell count.
Can be used to obtain absolute counts of cells or any other samples acquired on a flow cytometer and also suitable to work with all kinds of assays including a lyse, no-wash whole blood assay.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty