Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BioClean™ Clearview™ Autoclavable Thermoplastic Rubber Goggles, Ansell

Supplier: Ansell
BioClean Clearview™
BCAG BCAG
75833-997PK 3195.81 USD InStock
75833-997 CA75833-996
BioClean™ Clearview™ Autoclavable Thermoplastic Rubber Goggles, Ansell
Goggles
Optically correct for clear vision.

  • Super-soft frame
  • Upper vents and lower indirect ventilation system
  • Toughened polycarbonate, anti-scratch, and optically correct lens
  • Non-linting, latex-free head band
  • Tested to withstand over 40 autoclave cycles of 30 minutes duration at 121 °C (250 °F)
  • Can be worn over eye-glasses
  • Supplied non sterile
  • Can be autoclaved at 121 °C/250 °F for 30 minutes

BioClean™ Clearview™ autoclavable cleanroom goggles have a super-soft thermoplastic rubber frame with upper vents and an indirect lower ventilation system to provide the wearer comfort and reduce the risk of contamination entering a controlled environment. These goggles feature a toughened anti-scratch, optically clear polycarbonate lens for clear vision.

Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications and processed in an NEBB certified ISO Class 4 cleanroom environment.

Certifications: Conforms to Category 2 – Personal Protective Equipment PPE Directive 89/686/EEC. Complies with requirements of EN 166:2001 standard for personal eye protection.

Accessories information: The replacement lens for these autoclavable goggles is manufactured from toughened polycarbonate, and is optically correct and autoclavable. Replacement strap also available separately.

Packaging: One piece per sealed PE bag (to be removed prior to autoclaving); 12 bags per inner box; five boxes per lined carton (60 pieces). PLEASE NOTE: Product sold per inner box (12 pieces).

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty