Would you like to log in again?
BioClean™ Legion™ Non sterile Latex Gloves, Ansell
Supplier: Nitritex
BioClean Legion™
BLA301-S BLA302-M BLA304-XL BLA303-L
75834-010CS 771.6 USD InStock
75834-010 75834-012 75834-014 75834-008
BioClean™ Legion™ Non sterile Latex Gloves, Ansell
Gloves
BioClean™ Legion™ non-sterile latex gloves offer unbeatable comfort with elbow length protection.
  • Exceptional flexibility and comfort
  • Elbow length
  • Powder-free
  • Beaded cuff
  • Textured for improved grip
  • Double-donnable
  • ISO Class 5 compatible
  • Non-particulating EasyTear™ packaging
For supreme comfort, exceptional flexibility, and elbow length protection, the 40.6 cm (16”) BioClean™ Legion™ Latex Glove is the optimal choice. With a textured surface for enhanced grip, this ambidextrous natural-colour glove is powder-free and contains a latex protein level of less than 50 μg/g.
Certifications: Manufactured in a facility holding ISO 9001: 2008 and 89/686/EEC Article 11b certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC.
Processed in an NEBB certified ISO Class 5 environment.
Complies with the requirements of Glove Standard EN 420:2003 and EN 374-1, 2 & 3:2003.
Physical properties comply with European Medical Glove Standard EN455-2:2009.
Packaging: Gloves are packaged 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty