Would you like to log in again?
BioClean™ Nerva™ Non sterile Nitrile Gloves, Ansell
Supplier: Nitritex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BioClean™ BioClean Nerva™
BNAL-L BNAL-L BNAL-XL BNAL-M BNAL-S BNAL-XXL BNAL-S BNAL-XXL BNAL-XL BNAL-M
75834-040CS 862.68 USD InStock
75834-040 75834-042 CA75834-040 75834-044 CA75834-042 CA75834-044 75834-036 CA75834-036 75834-038 CA75834-038
BioClean™ Nerva™ Non sterile Nitrile Gloves, Ansell
Gloves
BioClean™ Nerva™ non-sterile nitrile gloves offer excellent comfort and elbow length protection.
  • Elbow length protection
  • Textured for improved grip
  • Non-particulating EasyTear™ packaging
With excellent ESD properties and latex-free construction, these ambidextrous gloves feature improved double-donning properties, enhanced grip, and a beaded cuff for added strength. Available in a blue or white finish.
Certifications: Manufactured in a facility holding ISO 9001: 2008.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 and EN 374-1, 2 & 3:2003.
Physical properties comply with European Medical Glove Standard EN 455-2:2009.
Packaging: Gloves are packaged 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces).These parts are set up as Case of 500 = 500 pairs = 1000 pieces
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty