Would you like to log in again?
BioClean™ Nitramax™ Sterile Nitril Gloves, Ansell
Supplier: Nitritex
BioClean Nitramax™ BioClean™
BNMS-100 BNMS-75 BNMS-100 BNMS-75 BNMS-90 BNMS-85 BNMS-65 BNMS-80 BNMS-70 BNMS-70 BNMS-65 BNMS-85 BNMS-60 BNMS-80 BNMS-90
75834-046CS 3190.32 USD InStock
CA75834-108 75834-046 CA75834-110 75834-112 CA75834-046 CA75834-112 75834-114 75834-104 CA75834-114 75834-116 75834-106 CA75834-116 CA75834-106 75834-108 75834-110
BioClean™ Nitramax™ Sterile Nitril Gloves, Ansell
Gloves
BioClean™ Nitramax™ Sterile Nitril Gloves offer full arm protection.
  • Chemical resistant
  • Textured for improved grip
  • Non-particulating EasyTear™ packaging
BioClean Nitramax™ Glove is specifically designed for use when full arm protection is required. These nitrile gloves provide exceptional comfort and flexibility, have excellent antistatic properties, and are resistant to a wide range of chemicals.
Certifications: Manufactured in a facility holding ISO 9001: 2008 and 89/686/EEC Article 11b certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of Glove Standard EN420:2003 and EN374-1, 2 & 3:2003.
Physical properties comply with European Medical Glove Standard EN455-2:2009.
Sterilized in accordance with ISO 11137-1:2006.
Packaging: Gloves are packaged one pair per inner PE wallet; one wallet per sealed EasyTear™ PE pouch; 10 pouches per sealed outer PE bag; 10 outer bags per lined carton (100 pairs).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty