Would you like to log in again?
EASY PURCHASE PACK P-SL 12X32MM 11MM 2ML
Supplier: MicroSolv Technology Corp
Easy Purchase Pack 2ml^ Easy Purchase Pack 2ml, 11mm clear glass snap/crimp top autosampler vials (12x32mm) with write on patch, and blue Soft-Guard snap caps with fitted preslit silicone/PTFE septa
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EASY PURCHASE PACK P-SL 12X32MM 11MM 2ML 75834-164 9502C-S-3WCP Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty