Would you like to log in again?
CAP AUTO SAMP AQ SCREW TOP 10B LT BL 9MM
Supplier: MicroSolv Technology Corp
Cap AQ* autosampler^ Caps, AQ* autosampler, screw top, dark blue with bonded Ultra Low Bleed silicone/PTFE septa. For use with 9-425mm, 12x32mm AQ vials
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP AUTO SAMP AQ SCREW TOP 10B LT BL 9MM 75834-170 9509S-10B Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty