Would you like to log in again?
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Supplier: MicroSolv
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
RSA-Pro™ AQ™ RSA™
9509S-3WCPEARS 9509S-1CP-RS 9532C-3CP-RS 9532S-1CP-RS 9512C-1MP-RS 9512S-1MP-RS 9509S-3WAP-RS 9512S-1AP-RS 9509C-1EWCP-RS 9509S-WDCV-MR 9512S-9CP-T-RS 9509C-3XWCP-RS 9532S-0DCV 9512S-1CP-RS 9509S-1WCPRSA 9532S-0CV-RS 9512S-1EMP-RS 9509C-1XWCP-RS 9532C-1CP-RS 9509S-0CV-RS 9512S-3CP-RS 9509S-WAV-RS 9532S-WCV-WT 9512C-0CV-T-RS 9509S-9WAP-RS 9509C-1WAP-RS 9532S-WCV 9532SWCVWTRS 9509C-3XWAP-RS 9512S-0AV-RS 9512S-9AP-RS 9512C-3MP-RS 9509S-WDAV-MR 9509S-3CP-RS 9532S-3DCP-MR 9509S-3WCP-RS 9509C-3EWCP-RS 9509S-3EWAP-RS 9532S1CPWTRS 9534S-0CV 9509S-3XWCP-RS 9532S3CPWTRS 9509S-1WAP-RS 9512S-0CV-RS 9509C-1WCP-RS 9512C-0CV-RS 9509S-3EWCP-RS 9512S-3AP-RS 9509C-WAV-RS 9509C-3WAP-RS 9509S-WCV-RS 9509S-1EWAP-RS 9512S-1ECP-RS 9532S-0AV-RS 9512S-9CP-RS 9509S-1XWAP-RS 9512C-1CP-RS 9512S-0CV-T-RS 9509S-1EWCP-RS 9532C-0CV-RS 9509S-1XWCP-RS 9509S-1WCP-RS 9532S-3CP-RS 9509C-WCV-RS 9509S-3XWAP-RS 9512C-3CP-RS 9509C-3WCP-RS 9509S-9WCP-RS 9512S-3MP-RS
76291-908PK 275.82 USD InStock
76291-908 89220-110 97056-760 97056-766 97056-764 97056-762 97057-130 30617-982 97055-982 97058-834 97055-984 10804-802 10804-804 97055-986 89220-128 89220-126 97057-178 89220-088 75856-552 89220-086 89220-120 97055-980 75834-182 97056-770 89220-080 10804-890 76057-634 75846-068 10804-808 76318-658 75834-180 97056-772 97055-454 10804-814 10804-816 97058-828 75846-066 97057-820 75834-168 89220-058 10804-896 97055-450 89220-130 10804-810 89220-098 75834-172 97057-818 89220-094 75834-174 89220-092 75834-176 76307-376 30618-018 97058-832 97052-682 97058-830 97052-680 10804-822 97057-274 10804-904 89220-108 89220-100 10804-820 89220-062 89220-060 75834-166 30621-376 10141-548 30621-374
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Vials
Reduced surface activity, non treated clear and amber glass vials. Convenient kits are packaged with high purity caps for low adsorbance.
  • Prevent pH changes in samples with water diluent
  • Great for bio active compounds
  • Minimize hydrophilic sample adsorption to the glass which is especially important for low abundance analytes
  • Prevent adducts seen in LCMS from borosilicate glass vials
  • Cleanest vials on the market
  • Superior dimensional control for compatibility with the most sensitive autosamplers
RSA™ autosampler vials prevent sample diluents (such as water) from de-protonating the many hydroxyl groups on the vial surface producing a negative charge on the glass. This 'acidic' functionality of these silanols has been known to cause adsorption of basic compounds, and/or hydrolyze some susceptible compounds making them undetectable. The very low surface metals in these vials prevent contamination, precipitation, chelation or cause other changes in your samples as with standard borosilicate glass. These vials are chosen for very low abundance analytes such as low dosage form pharmaceuticals, forensic samples, bio-markers, environmental samples and other critical low concentration samples.
Packaging: RSA™ easy purchase packs are packaged in a clam shell container with two individual compartments that hold 100 vials and 100 caps.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty