Would you like to log in again?
508.1 Calibration Mix #1 (17 Components), Restek
Supplier: Restek
32094
75834-222EA 108.64 USD InStock
75834-222
508.1 Calibration Mix #1 (17 Components), Restek
Standards Organic Standards
Consists of 17 components, those are aldrin, α-bhc, β-bhc, δ-bhc, γ-bhc , 4,4'-ddd, 4,4'-dde, 4,4'-ddt, dieldrin, endosulfan i, endosulfan ii, endosulfan sulfate, endrin, endrin aldehyde, heptachlor, heptachlor epoxide and methoxychlor.
17 components
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty