Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD86 Rat Monoclonal Antibody (PE-Cyanine7) [clone: GL-1 (GL1)]

Supplier: Tonbo Biosciences
60-0862-U100 60-0862-U025
75834-282EA 85.6 USD InStock
75834-282 75834-284
Anti-CD86 Rat Monoclonal Antibody (PE-Cyanine7) [clone: GL-1 (GL1)]
Antibodies
The GL-1 antibody reacts with mouse CD86, also known as B7-2, an 80 kDa cell surface protein which is a ligand for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 can also bind a second B7 ligand known as CD80 (B7-1). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response. The GL-1 antibody may be used as a marker for CD86 expression on B cells, macrophages, and dendritic cells.

Type: Primary
Antigen: CD86 (B7-2)
Clonality: Monoclonal
Clone: GL-1 (GL1)
Conjugation: PE-Cyanine7
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty