Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HLA-DR Mouse Monoclonal Antibody (PerCP) [clone: L243]

Supplier: Tonbo Biosciences
67-9952-T025 67-9952-T100
75834-314EA 145 USD InStock
75834-314 75834-316
Anti-HLA-DR Mouse Monoclonal Antibody (PerCP) [clone: L243]
Antibodies
The L243 antibody reacts with a member of the human MHC Class II antigens, HLA-DR. The HLA-DR antigen is expressed on B lymphocytes, activated T lymphocytes, activated NK cells, monocytes, macrophages, other antigen presenting cells and progenitor cells. The L243 antibody is specific to an epitope on the alpha subunit of the heterodimeric HLA-DR protein and binds a different epitope than the LN3 antibody clone. It does not cross-react with HLA-DP or HLA-DQ. This antibody is reported to be cross-reactive with non-human primates including Chimpanzee, Cynomolgus, Rhesus and Baboon. Please choose the appropriate format for each application.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: L243
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty