Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD20 Mouse Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 2H7]

Supplier: Tonbo Biosciences
65-0209-T025 65-0209-T100
75834-360EA 381 USD InStock
75834-360 75834-358
Anti-CD20 Mouse Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 2H7]
Antibodies
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation. The 2H7 antibody is reported to be cross-reactive with several non-human species including Baboon, Chimpanzee, Cynomolgus and Rhesus. Please choose the appropriate format for each application.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: PerCP-Cyanine5.5
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty