Would you like to log in again?
OLC 03.2 VOA MegaMix Standard, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
30492
75834-466EA 66.93 USD InStock
75834-466
OLC 03.2 VOA MegaMix Standard, Restek
Standards Organic Standards
VOA MegaMix (with m- and p-xylene isomers at half the concentration of other components) is a deuterated monitoring compounds (DMCs) mix, a fortification mix for all compounds requiring calibration at higher concentration, and single-compound solutions of 3,3'-dichlorobenzidine and 4-chloroaniline.
42 components
Components: Benzene, bromochloromethane, bromodichloromethane, bromoform, carbondisulphide, carbontetrachloride, chlorobenzene, chloroform, cyclohexane, dibromochloromethane (chlorodibromomethane), 1,2-dibromo-3-chloropropane (DBCP), 1,2-dibromoethane (EDB), 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, 1,1-dichloroethane, 1,2-dichloroethane, 1,1-dichloroethene, cis-1,2-dichloroethene, trans-1,2-dichloroethene, 1,2-dichloropropane, cis-1,3-dichloropropene, trans-1,3-dichloropropene, ethylbenzene, isopropylbenzene (cumene), methyl acetate, methylcyclohexane, methyl tert-butyl ether (MTBE), methylene chloride (dichloromethane), styrene, 1,1,2,2-tetrachloroethane, tetrachloroethene, toluene, 1,2,3-trichlorobenzene, 1,2,4-trichlorobenzene, 1,1,1-trichloroethane, 1,1,2-trichloroethane, trichloroethene, 1,1,2-trichlorotrifl uoroethane (CFC-113), m-xylene, o-xylene and p-xylene.
The concentration of m-xylene and p-xylene is 1000 µg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty