Would you like to log in again?
Acrolein/Acrylonitrile, Restek
Supplier: Restek
30600
75834-498EA 46.56 USD InStock
75834-498
Acrolein/Acrylonitrile, Restek
Standards Organic Standards
2 components
Ordering information: Must ship overnight on ice.
This product has a limited shelf life. We recommend that you order only the ampul quantity that meets your immediate needs.
NOTE: Store standards under frozen conditions (0 °C or colder). We recommend you store ampuls lying flat (horizontally) to prevent breakage. For best results, thaw the contents of the ampul fully and shake the ampul well before opening and using the solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty