Would you like to log in again?
DHA Naphthenes Standard, Restek
Supplier: Restek
30728
75834-584EA 430.13 USD InStock
75834-584
DHA Naphthenes Standard, Restek
Standards Organic Standards
ASTM Methods D6729, D6730, and D6733 (Determination of Individual Components in Spark Ignition Engine Fuels) are designed for the determination of the individual hydrocarbons present in spark ignition engine fuels, as well as fuel blends containing oxygenates such as methyl tert-butyl ether, ethyl tert-butyl ether, tert-butanol, ethanol, etc.
This standard is produced for refineries performing Detailed Hydrocarbon Analysis (DHA) of crude oil feedstocks and fuels. It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for quantitative calibration to help our customers optimize production and maximize profitability—while fulfilling requirements such as ASTM Methods D6729, D6730, and D6733.
26 components
Certifications: ASTM Methods D6729, D6730, and D6733 (Determination of Individual Components in Spark Ignition Engine Fuels)
Ordering information: Quantity discounts not available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty