Would you like to log in again?
DHA Aromatics Standard (37 Components), Restek
Supplier: Restek
30729
75834-586EA 430.13 USD InStock
75834-586
DHA Aromatics Standard (37 Components), Restek
Standards Organic Standards
This standard is produced for refineries performing Detailed Hydrocarbon Analysis (DHA) of crude oil feedstocks and fuels.
This standard contain 37 components: Benzene (8,255%), N-Butylbenzene (2,077%), sec-Butylbenzene (3,09%), tert-Butylbenzene (2,058%), tert-1-Butyl-4-Ethylbenzene (2,076%), tert-1-Butyl-2-Methylbenzene (2,077%), tert-1-Butyl-3,4,5-Trimethylbenzene (3,355%), 1,2-Dimethyl-3-Ethylbenzene (2,152%), 1,2-Dimethyl-4-Ethylbenzene (0,915%), 1,3-Dimethyl-2-Ethylbenzene (0,799%), 1,3-Dimethyl-5-Ethylbenzene (0,407%), 1,4-Dimethyl-2-Ethylbenzene (2,076%), Ethylbenzene (8,248%), Hexylbenzene (5,155%), Isobutylbenzene (3,099%), Isopropylbenzene (2,085%), 2-Methylbutylbenzene (0,118%).
It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for quantitative calibration.
Certifications: Meets requirements such as ASTM Methods D6729, D6730, and D6733. Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: Quantity discounts not available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty