Would you like to log in again?
Base/Neutrals Matrix Spike Standards, Restek
Supplier: Restek
31004-15 31074 31004 31084
RK31084EA 120.28 USD InStock
RK31084 75834-594 RK31074 RK31004
Base/Neutrals Matrix Spike Standards, Restek
Standards Environmental Analysis Standards
Standards are packaged in methanol in an ampule.
  • Highest concentrations commercially available
  • Reduces laboratory cost per sample extract
  • Mixture contains Acenaphthene, 1,4-Dichlorobenzene, 2,4-Dinitrotoluene, N-Nitroso-di-n-propylamine, Pyrene, and 1,2,4-Trichlorobenzene
Each standard includes an ampule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty