Would you like to log in again?
610 PAH Calibration Mix B (16 Components), Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
31455
75834-700EA 64.99 USD InStock
75834-700
610 PAH Calibration Mix B (16 Components), Restek
Standards Organic Standards
This 610 PAH calibration mix B idela for LC-UV detection.
It consists of 16 components: Acenaphthene (1000 μg/ml), Acenaphthylene (2000 μg/ml), Anthracene(100 μg/ml), Benz(a)anthracene (100 μg/ml), Benzo(a)pyrene (100 μg/ml), Benzo(b)fluoranthene (200 μg/ml), Benzo(k)fluoranthene (100 μg/ml), Benzo(ghi)perylene (200 μg/ml), Chrysene (100 μg/ml), Dibenz(a,h)anthracene (200 μg/ml), Fluoranthene (200 μg/ml), Fluorene (200 μg/ml), Indeno(1,2,3-cd)pyrene (100 μg/ml), Naphthalene (1000 μg/ml), Phenanthrene (100 μg/ml) and Pyrene (100 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty